2-(Tributylstannyl)thiophene ≥97%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
H55904.AA
H55904.AC
H55904.AE
H55904.AAEA
1410
CZK
H55904.AA
H55904.AC
H55904.AE
2-(Tributylstannyl)thiophene ≥97%
2-(Tributylstannyl)thiophene
Vzorec:
C₁₆H₃₀SSn Bod varu: 155 °C (0,1 mmHg) Hustota: 1,175 Storage Temperature: Ambient |
MDL Number:
MFCD00192513 CAS číslo: 54663-78-4 UN: 2788 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...