5-Chloro-1-(4-piperidinyl)-2-benzimidazolidinone ≥98%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
5-Chloro-1-(piperidin-4-yl)-1H-benzo[d]imidazol-2(3H)-one, 5-Chloro-1-(4-piperidinyl)-2-benzimidazolinone
B22521.18
B22521.09
B22521.30
B22521.18EA
6460
CZK
B22521.18
B22521.09
B22521.30
5-Chloro-1-(4-piperidinyl)-2-benzimidazolidinone ≥98%
5-Chloro-1-(4-piperidinyl)-2-benzimidazolidinone
Purity: 98%
CAS No.: 53786-28-0
Molecular Formula: C12H14ClN3O
Formula Weight: 251.71
Melting Point: 227-230°
MDL No.: MFCD02684518
Vzorec:
C₁₂H₁₄ClN₃O MW: 251.72 g/mol Bod tání: 227…230 °C Storage Temperature: Ambient |
MDL Number:
MFCD02093793 CAS číslo: 53786-28-0 EINECS: 258-771-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...