4-(Morpholino)phenylboronic acid pinacol ester 95%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
4-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]morpholine, 4-Morpholinophenylboronic acid Pinacol ester, 4-(4-Morpholinyl)benzeneboronic acid pinacol ester, 4-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)morpholine
H51925.03
H51925.06
H51925.03EA
2260
CZK
H51925.03
H51925.06
4-(Morpholino)phenylboronic acid pinacol ester 95%
4-(Morpholino)phenylboronic acid pinacol ester
Purity: 95%
CAS No.: 568577-88-8
Molecular Formula: C16H24BNO3
Formula Weight: 289.18
MDL No.: MFCD04112544
Vzorec:
C₁₆H₂₄BNO₃ MW: 289,18 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD04112544 CAS číslo: 568577-88-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...