Tisk…

3-(4-Methyl-1-piperazinylcarbonyl)phenylboronic acid pinacol ester ≥97%

Dodavatel: Thermo Fisher Scientific
Varování

H29224.06 H29224.03
H29224.06EA 7040 CZK
H29224.06 H29224.03
3-(4-Methyl-1-piperazinylcarbonyl)phenylboronic acid pinacol ester ≥97%
3-(4-Methyl-1-piperazinylcarbonyl)phenylboronic acid pinacol ester

Purity: 97% CAS No.: 883738-38-3 Molecular Formula: C18H27BN2O3 Formula Weight: 330.23 Melting Point: 86-89° MDL No.: MFCD05864308

Vzorec: C₁₈H₂₇BN₂O₃
Bod tání: 86…89 °C
Storage Temperature: Ambient
MDL Number: MFCD05864308
CAS číslo: 883738-38-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR