4-(4-Morpholinylcarbonyl)phenylboronic acid ≥98%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
H53126.MD
H53126.03
H53126.MDEA
2270
CZK
H53126.MD
H53126.03
4-(4-Morpholinylcarbonyl)phenylboronic acid ≥98%
4-(4-Morpholinylcarbonyl)phenylboronic acid
Purity: 98%
CAS No.: 389621-84-5
Molecular Formula: C11H14BNO4
Formula Weight: 235.05
Melting Point: 126-132°
MDL No.: MFCD03411952
Vzorec:
C₁₁H₁₄BNO₄ Bod tání: 126…132 °C Storage Temperature: Ambient |
MDL Number:
MFCD03411952 CAS číslo: 389621-84-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...