Uracil-5-boronic acid 95%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
2,4-Dihydroxypyrimidine-5-boronic acid, (2,4-Dioxo-1,2,3,4-tetrahydropyrimidin-5-yl)boronic acid
H27219.MD
H27219.03
H27219.MDEA
6040
CZK
H27219.MD
H27219.03
Uracil-5-boronic acid 95%
Uracil-5-boronic acid
Purity: 95%
CAS No.: 70523-22-7
Molecular Formula: C4H5BN2O4
Formula Weight: 155.90
Storage Temperature: -30°C to -10°C
Melting Point:
Vzorec:
C₄H₅BN₂O₄ MW: 155.90 g/mol |
MDL Number:
MFCD01318983 CAS číslo: 70523-22-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...