Tisk…

4-Chloro-N-methylpicolinamide ≥97%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: 4-Chloro-N-methylpyridine-2-carboxamide

H50535.03 H50535.MD
H50535.03EA 7310 CZK
H50535.03 H50535.MD
4-Chloro-N-methylpicolinamide ≥97%
4-Chloro-N-methylpicolinamide

Purity: 97% CAS No.: 220000-87-3 Molecular Formula: C7H7ClN2O Formula Weight: 170.60 Melting Point: 39-43° MDL No.: MFCD02185921

Vzorec: C₇H₇ClN₂O
MW: 170.60 g/mol
Bod tání: 39…43 °C
Storage Temperature: Ambient
MDL Number: MFCD02185921
CAS číslo: 220000-87-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR