Tisk…

Tartronic acid ≥98%

Dodavatel: Thermo Fisher Scientific
Varování

B21640.02 B21640.77
B21640.02EA 4740 CZK
B21640.02 B21640.77
Tartronic acid ≥98%
Tartronic acid

Purity: 98% CAS No.: 80-69-3 Molecular Formula: C3H4O5 Formula Weight: 120.06 Physical Form: Powder Appearance: White Melting Point: ca 151° dec. Beilstein Reference: 1209791 Merck Reference: 14,9073 MDL No.: MFCD00004237

Vzorec: C₃H₄O₅
Storage Temperature: Ambient
MDL Number: MFCD00004237
CAS číslo: 80-69-3
EINECS: 201-301-1
Merck Index: 14,9073

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR