Tisk…

4-(Benzyloxy)aniline hydrochloride, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 4-Benzyloxyanilinium Chloride, 4-Aminophenyl benzyl ether hydrochloride, p-Benzyloxyaniline hydrochloride

M-Clarity™ quality level = MQ100

116637-100G 116637-25G
SIAL116637-100GEA 4720 CZK
SIAL116637-100G SIAL116637-25G
4-(Benzyloxy)aniline hydrochloride, Sigma-Aldrich®
4-(Benzyloxy)aniline hydrochloride
Vzorec: C₁₃H₁₄ClNO
MW: 235,711 g/mol
MDL Number: MFCD00012995
CAS číslo: 51388-20-6
EINECS: 257-170-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR