Tisk…

tert-Butyl-4-(bromomethyl)benzoate 95%

Dodavatel: Thermo Scientific

Synonyms: 4-Bromo-methyl-benzoic acid mono tert-butyl ester, tert-Butyl 4-(bromomethyl)benzoate

H35540.03 H35540.09
H35540.03EA 2270 CZK
H35540.03 H35540.09
tert-Butyl-4-(bromomethyl)benzoate 95%
tert-Butyl-4-(bromomethyl)benzoate
Vzorec: C₁₂H₁₅BrO₂
MW: 271,15 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD04973450
CAS číslo: 108052-76-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR