Tisk…

Probenecid

Dodavatel: Cayman Chemical

14981-5 14981-10 14981-25 14981-100
CAYM14981-5EA 1320 CZK
CAYM14981-5 CAYM14981-10 CAYM14981-25 CAYM14981-100
Probenecid
Probenecid
Vzorec: C₁₃H₁₉NO₄S
MDL Number: MFCD00038402
CAS číslo: 57-66-9
EINECS: 200-344-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR