α-Bromo-4-chlorophenylacetic acid ≥97%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
H60141.03
H60141.06
H60141.03EA
1330
CZK
H60141.03
H60141.06
α-Bromo-4-chlorophenylacetic acid ≥97%
α-Bromo-4-chlorophenylacetic acid
Purity: 97%
CAS No.: 3381-73-5
Molecular Formula: C8H6BrClO2
Formula Weight: 249.49
Melting Point: 95-99°
UN No.: 3261
MDL No.: MFCD08276760
Vzorec:
C₈H₆BrClO₂ MW: 249,49 g/mol Bod tání: 95…99 °C Storage Temperature: Ambient |
MDL Number:
MFCD08276760 CAS číslo: 3381-73-5 UN: 3261 ADR: 8,II |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...