1-[(Ammoniooxy)methyl]-4-methoxybenzene chloride 98%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
O-(4-Methoxybenzyl)hydroxylamine hydrochloride
H63312.03
H63312.06
H63312.03EA
2480
CZK
H63312.03
H63312.06
1-[(Ammoniooxy)methyl]-4-methoxybenzene chloride 98%
1-[(Ammoniooxy)methyl]-4-methoxybenzene chloride
Purity: 98%
CAS No.: 21038-22-2
Molecular Formula: C8H11NO2
Formula Weight: 153.18
Physical Form: Solid
Appearance: White
MDL No.: MFCD01114582
Vzorec:
C₈H₁₂ClNO₂ MW: 189,64 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01114582 CAS číslo: 876-33-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...