Tisk…

2-Thiopheneboronic acid pinacol ester ≥98%

Dodavatel: Thermo Fisher Scientific

H63819.03 H63819.06 H63819.14
H63819.03EA 1350 CZK
H63819.03 H63819.06 H63819.14
2-Thiopheneboronic acid pinacol ester ≥98%
2-Thiopheneboronic acid pinacol ester

Purity: 98% CAS No.: 193978-23-3 Molecular Formula: C10H17BO3S Formula Weight: 228.12 Storage Temperature: -30°C to -10°C Physical Form: Solid Melting Point: 65-70° MDL No.: MFCD05663878

Vzorec: C₁₀H₁₅BO₂S
Bod tání: 65…70 °C
Storage Temperature: Freezer
MDL Number: MFCD05663878
CAS číslo: 193978-23-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR