Tisk…

4-Fluoro-3-nitroanisole 95%

Dodavatel: Thermo Scientific
Varování

Synonyms: 1-Fluoro-4-methoxy-2-nitrobenzene

H61554.03 H61554.06
H61554.03EA 4090 CZK
H61554.03 H61554.06
4-Fluoro-3-nitroanisole 95%
4-Fluoro-3-nitroanisole
Vzorec: C₇H₆FNO₃
MW: 171,13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11848442
CAS číslo: 61324-93-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR