Tisk…

4-Biphenylboronic acid pinacol ester ≥97%

Dodavatel: Thermo Fisher Scientific

H59919.03 H59919.06
H59919.03EA 2110 CZK
H59919.03 H59919.06
4-Biphenylboronic acid pinacol ester ≥97%
4-Biphenylboronic acid pinacol ester
Vzorec: C₁₈H₂₁BO₂
MW: 280.17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07368291
CAS číslo: 144432-80-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR