2-Fluoro-4-iodobenzonitrile ≥98%
Dodavatel: Thermo Scientific
|
Varování
|
H61973.03
H61973.06
H61973.03EA
2720
CZK
H61973.03
H61973.06
2-Fluoro-4-iodobenzonitrile ≥98%
2-Fluoro-4-iodobenzonitrile
Purity: 98%
CAS No.: 137553-42-5
Molecular Formula: C7H3FIN
Formula Weight: 247.01
Physical Form: Powder
Sensitivity: Light Sensitive
Appearance: Red-brown
UN No.: 3439
MDL No.: MFCD03094172
Vzorec:
C₇H₃FIN MW: 247,01 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03094172 CAS číslo: 137553-42-5 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...