2-Amino-3-fluorobenzonitrile 95%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
H61580.MD
H61580.03
H61580.MDEA
1530
CZK
H61580.MD
H61580.03
2-Amino-3-fluorobenzonitrile 95%
2-Amino-3-fluorobenzonitrile
Vzorec:
C₇H₅FN₂ Storage Temperature: Ambient |
MDL Number:
MFCD07368774 CAS číslo: 115661-37-5 UN: 3439 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...