Pinostilbene
Dodavatel: Cayman Chemical
Synonyms:
(E)-3-(4-Hydroxystyryl)-5-methoxyphenol
14044-1
14044-5
14044-10
14044-25
CAYM14044-1EA
1050
CZK
CAYM14044-1
CAYM14044-5
CAYM14044-10
CAYM14044-25
Pinostilbene
Pinostilbene
Vzorec:
C₁₅H₁₄O₃ |
MDL Number:
MFCD12407153 CAS číslo: 42438-89-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...