Tisk…
Plunger cans
Dodavatel: asecos
23133 23134 23135 23136 23137 23138 23139 40097 40098
MSGA23133EA 2230 CZK
MSGA23133 MSGA23134 MSGA23135 MSGA23136 MSGA23137 MSGA23138 MSGA23139 MSGA40097 MSGA40098
Plunger cans
Nádoby bezpečnostní
These plunger cans are dedicated to the safe moistening of cloth, wipes and tissues.
  • Unused liquid flows back into the container
  • Spring mounted plate includes flame barrier
  • Shock resistant and explosion proof (PE models only)
  • Cans with protector ring (stainless steel models only)