Tisk…
ESD safety shoes, lace-up, HACCP, Anatom, 32131/32136
Dodavatel: Abeba
32136/43 32131/36 32131/37 32131/38 32131/39 32131/40 32131/41 32131/42 32131/43 32131/44 32131/45 32131/46 32131/47 32131/48 32136/36 32136/37 32136/38 32136/39 32136/40 32136/41 32136/42 32136/44 32136/45 32136/46 32136/47 32136/48 32131/49 32136/49 32136/50 32131/50
ABEB32136/43EA 3530 CZK
ABEB32136/43 ABEB32131/36 ABEB32131/37 ABEB32131/38 ABEB32131/39 ABEB32131/40 ABEB32131/41 ABEB32131/42 ABEB32131/43 ABEB32131/44 ABEB32131/45 ABEB32131/46 ABEB32131/47 ABEB32131/48 ABEB32136/36 ABEB32136/37 ABEB32136/38 ABEB32136/39 ABEB32136/40 ABEB32136/41 ABEB32136/42 ABEB32136/44 ABEB32136/45 ABEB32136/46 ABEB32136/47 ABEB32136/48 ABEB32131/49 ABEB32136/49 ABEB32136/50 ABEB32131/50
ESD safety shoes, lace-up, HACCP, Anatom, 32131/32136
Obuv
ESD shoe made from microfibre upper and a fuel resistant PU outsole with high slip resistance and a self-cleaning profile.
  • Perspiration-absorbent special lining with silver fibre
  • Replaceable acc Wave insole; orthopaedic insoles possible
  • Asymmetric heel and Flexzone for a natural bedding and foot movement
  • TPU torsion waist, arch support and high stability
  • Shock-absorbent PU midsole
  • Sanitized® treatment reduces odour and bacterial growth
  • Protective steel toecap (200 J)
  • Washable at 30 °C
Suitable for hygienically demanding areas.
Colour: White or black
Certifikace: EN 61340 (ESD); DGUV 112-191; HACCP; EN ISO 20345:2011; S2, SRC