Tisk…

Ytterbium(III) sulphate octahydrate ≥99.9% (REO, rare earth oxide basis), REacton®

Dodavatel: Thermo Scientific
Varování

Synonyms: Ytterbium(III) sulphate octahydrate

14572.09 14572.18
14572.09EA 1870 CZK
14572.09 14572.18
Ytterbium(III) sulphate octahydrate ≥99.9% (REO, rare earth oxide basis), REacton®
Ytterbium(III) sulphate octahydrate

Purity: 99.9% CAS No.: 10034-98-7 Molecular Formula: Yb2(SO4)3.8H2O Formula Weight: 778.38 (634.26anhy) Physical Form: Crystalline

Vzorec: O₁₂S₃Yb₂·8H₂O
MW: 778,396 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00149887
CAS číslo: 10034-98-7
EINECS: 236-727-7
Merck Index: 14,10105

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR