Tisk…

Gadolinium(III) sulphate octahydrate ≥99.9% (REO, rare earth oxide basis), REacton®

Dodavatel: Thermo Scientific
Varování

42869.09 42869.18
42869.09EA 3010 CZK
42869.09 42869.18
Gadolinium(III) sulphate octahydrate ≥99.9% (REO, rare earth oxide basis), REacton®
Gadolinium(III) sulphate octahydrate

Purity: 99.9% CAS No.: 13450-87-8 Molecular Formula: Gd2(SO4)3.8H2O Formula Weight: 746.81 (602.68anhy) Physical Form: Crystalline Aggregates Density: 4.139 Solubility: Soluble in cold water Merck Reference: 14,4326 MDL No.: MFCD00149734


Cryogenic research

Vzorec: Gd₂O₁₂S₃·8H₂O
MW: 746,81 g/mol
Hustota: 4,139
Storage Temperature: Ambient
MDL Number: MFCD00149734
CAS číslo: 13450-87-8
EINECS: 237-115-2
Merck Index: 14,4326

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR