Tisk…

Picolinamide, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 2-Pyridinecarboxamide, Pyridine-2-carboxamide

M-Clarity™ quality level = MQ100

104051-10G 104051-50G
SIAL104051-10GEA 3130 CZK
SIAL104051-10G SIAL104051-50G
Picolinamide, Sigma-Aldrich®
Picolinamide
Vzorec: C₆H₆N₂O
MW: 122,13 g/mol
MDL Number: MFCD00023483
CAS číslo: 1452-77-3
EINECS: 215-921-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR