Tisk…

Trimellitic acid tris(2-ethylhexyl) ester, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Totm, Tris(2-ethylhexyl) benzene-1,2,4-tricarboxylate, Kodaflex TOTM, Tri(2-ethylhexyl) trimellitate

M-Clarity™ quality level = MQ200

538140-1L 538140-4L
SIAL538140-1LEA 2800 CZK
SIAL538140-1L SIAL538140-4L
Trimellitic acid tris(2-ethylhexyl) ester, Sigma-Aldrich®
Trimellitic acid tris(2-ethylhexyl) ester
Vzorec: C₃₃H₅₄O₆
MDL Number: MFCD00053799
CAS číslo: 3319-31-1
EINECS: 222-020-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR