Tisk…

2-Mercapto-1,3,4-thiadiazole ≥98%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: 1,3,4-Thiadiazole-2-thiol

B22556.09 B22556.18
B22556.09EA 1150 CZK
B22556.09 B22556.18
2-Mercapto-1,3,4-thiadiazole ≥98%
2-Mercapto-1,3,4-thiadiazole

Purity: 98% CAS No.: 18686-82-3 Molecular Formula: C2H2N2S2 Formula Weight: 118.17 Melting Point: 142-144° MDL No.: MFCD00040404

Vzorec: C₂H₂N₂S₂
MW: 118.17 g/mol
Bod tání: 142…144 °C
Storage Temperature: Ambient
MDL Number: MFCD00040404
CAS číslo: 18686-82-3
EINECS: 242-504-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR