Tisk…

Methyl-4-bromo-3-hydroxythiophene-2-carboxylate ≥97%

Dodavatel: Thermo Fisher Scientific

Synonyms: Methyl 4-bromo-3-hydroxythiophene-2-carboxylate

B20249.09 B20249.88
B20249.09EA 6940 CZK
B20249.09 B20249.88
Methyl-4-bromo-3-hydroxythiophene-2-carboxylate ≥97%
Methyl-4-bromo-3-hydroxythiophene-2-carboxylate

Purity: 97% CAS No.: 95201-93-7 Molecular Formula: C6H5BrO3S Formula Weight: 237.08 Melting Point: 79-80° MDL No.: MFCD00052081

Vzorec: C₆H₅BrO₃S
Bod tání: 79…80 °C
Storage Temperature: Ambient
MDL Number: MFCD00052081
CAS číslo: 95201-93-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR