Tisk…

Ethyl-5-bromothiophene-2-carboxylate ≥99%

Dodavatel: Thermo Scientific
Varování

Synonyms: Ethyl 5-bromothiophene-2-carboxylate

B22355.06 B22355.03
B22355.06EA 3640 CZK
B22355.06 B22355.03
Ethyl-5-bromothiophene-2-carboxylate ≥99%
Ethyl-5-bromothiophene-2-carboxylate

Purity: 99% CAS No.: 5751-83-7 Molecular Formula: C7H7BrO2S Formula Weight: 235.10 Boiling Point: 94-96°/4mm MDL No.: MFCD02683089

Vzorec: C₇H₇BrO₂S
MW: 235,10 g/mol
Bod varu: 94…96 °C (4 mmHg)
Storage Temperature: Ambient
MDL Number: MFCD02683089
CAS číslo: 5751-83-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR