Tisk…

2-Formyl-6-picoline, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 6-Methylpyridine-2-carboxaldehyde, 6-Methyl-2-pyridinecarboxaldehyde

M-Clarity™ quality level = MQ100

M78208-25G M78208-5G
SIALM78208-25GEA 23950 CZK
SIALM78208-25G SIALM78208-5G
2-Formyl-6-picoline, Sigma-Aldrich®
2-Formyl-6-picoline
Vzorec: C₇H₇NO
MW: 121,14 g/mol
MDL Number: MFCD00006291
CAS číslo: 1122-72-1
EINECS: 214-359-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR