2-Naphthalenemethanol, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
187313-25G
187313-5G
SIAL187313-25GEA
12350
CZK
SIAL187313-25G
SIAL187313-5G
2-Naphthalenemethanol, Sigma-Aldrich®
2-Naphthalenemethanol
Vzorec:
C₁₁H₁₀O MW: 158.2000 g/mol |
MDL Number:
MFCD00004124 CAS číslo: 1592-38-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...