Tisk…
1,1'-Binaphthyl-2,2'-diyl hydrogen phosphate ≥99%
Dodavatel: Thermo Scientific
Synonyms: 1,1-Binaphthyl-2,2-diyl hydrogenphosphate
H60395.06 H60395.03
H60395.06EA 3570 CZK
H60395.06 H60395.03
1,1'-Binaphthyl-2,2'-diyl hydrogen phosphate ≥99%
1,1'-Binaphthyl-2,2'-diyl hydrogen phosphate
Vzorec: C₂₀H₁₃O₄P
MW: 348,29 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00010045
CAS číslo: 35193-63-6
EINECS: 252-425-8