Tisk…

1-(3-Chloro-2-pyridinyl)piperazine ≥98%

Dodavatel: Thermo Fisher Scientific
Varování

H50859.03 H50859.MD
H50859.03EA 7790 CZK
H50859.03 H50859.MD
1-(3-Chloro-2-pyridinyl)piperazine ≥98%
1-(3-Chloro-2-pyridinyl)piperazine

Purity: 98% CAS No.: 87394-55-6 Molecular Formula: C9H12ClN3 Formula Weight: 197.67 MDL No.: MFCD08061034

Vzorec: C₉H₁₂ClN₃
Storage Temperature: Ambient
MDL Number: MFCD08061034
CAS číslo: 87394-55-6

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR