3-(4-Chlorophenoxy)piperidine ≥97%
Dodavatel: Thermo Scientific
|
Varování
|
H50935.03
H50935.MD
H50935.03EA
6040
CZK
H50935.03
H50935.MD
3-(4-Chlorophenoxy)piperidine ≥97%
3-(4-Chlorophenoxy)piperidine
Purity: 97%
CAS No.: 384346-27-4
Molecular Formula: C11H14ClNO
Formula Weight: 211.69
MDL No.: MFCD08061075
Vzorec:
C₁₁H₁₄ClNO MW: 211,69 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD08061075 CAS číslo: 384346-27-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...