5-(2-Pyridyl)-1H-tetrazole ≥98%
Dodavatel: Thermo Scientific
Synonyms:
2-(1H-Tetrazol-5-yl)pyridine, 5-(2-Pyridyl)-1H-tetrazole
B20852.03
B20852.06
B20852.03EA
3540
CZK
B20852.03
B20852.06
5-(2-Pyridyl)-1H-tetrazole ≥98%
5-(2-Pyridyl)-1H-tetrazole
Purity: 98%
CAS No.: 33893-89-9
Molecular Formula: C6H5N5
Formula Weight: 147.14
MDL No.: MFCD00068114
Vzorec:
C₆H₅N₅ MW: 147,14 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00068114 CAS číslo: 33893-89-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...