4-(Tributylstannyl)thiazole 95%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
H51457.MD
H51457.03
H51457.MDEA
2630
CZK
H51457.MD
H51457.03
4-(Tributylstannyl)thiazole 95%
4-(Tributylstannyl)thiazole
Purity: 95%
CAS No.: 173979-01-6
Molecular Formula: C15H29NSSn
Formula Weight: 374.17
UN No.: 2788
MDL No.: MFCD01319073
Vzorec:
C₁₅H₂₉NSSn Storage Temperature: Ambient |
MDL Number:
MFCD01319073 CAS číslo: 173979-01-6 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...