6-Methoxy-2-(tributylstannyl)pyrimidine, Sigma-Aldrich®
Dodavatel: Merck
|
Nebezpečí
|
M-Clarity™ quality level = MQ100
707031-1G
707031-500MG
SIAL707031-1GEA
9900
CZK
SIAL707031-1G
SIAL707031-500MG
6-Methoxy-2-(tributylstannyl)pyrimidine, Sigma-Aldrich®
6-Methoxy-2-(tributylstannyl)pyrimidine
Vzorec:
C₁₇H₃₂N₂OSn |
MDL Number:
MFCD15143643 CAS číslo: 850501-35-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...