Tisk…

Bis(benzonitrile)dichloroplatinum(II) (≥40% Pt)

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: Bis(benzonitrile)platinum(II) chloride, Dichlorobis(benzonitrile)platinum(II)

10004.02 10004.03 10004.06
10004.02EA 3850 CZK
10004.02 10004.03 10004.06
Bis(benzonitrile)dichloroplatinum(II) (≥40% Pt)
Bis(benzonitrile)dichloroplatinum(II)

CAS No.: 14873-63-3 Molecular Formula: PtCl2[(C6H5)(CN)]2 Formula Weight: 472.25 Physical Form: Powder Solubility: Slightly soluble in acetone, chloroform UN No.: 3439 MDL No.: MFCD00013125

Vzorec: C₁₄H₁₀Cl₂N₂Pt
MW: 472.233 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00013125
CAS číslo: 14873-63-3
EINECS: 238-943-7
ADR: 6.1 III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR