Tisk…
Tungsten (gold coated) ≥99.95% (metals basis) Au 99.99%, wire, Ø 0.025 mm (0.001 in)
Dodavatel: Thermo Fisher Scientific
41038.G5 41038.G6
41038.G5EA 10920 CZK
41038.G5 41038.G6
Tungsten (gold coated) ≥99.95% (metals basis) Au 99.99%, wire, Ø 0.025 mm (0.001 in)
Tungsten (gold coated)
Purity: 99.95% Physical Form: ≈0.005g/m (W only), 3-5 wt% Au plating Odour: Odourless MDL No.: MFCD00011461
Microelectronics, evaporation source, high temperature
Vzorec: W
Storage Temperature: Ambient
MDL Number: MFCD00011461
CAS číslo: 7440-33-7
EINECS: 231-143-9