Tisk…
Tungsten (gold coated), gauze woven from wire 50 mesh woven from 0.025mm (0.001in)
Dodavatel: Thermo Fisher Scientific
40926.FL 40926.GT
40926.FLEA 12260 CZK
40926.FL 40926.GT
Tungsten (gold coated), gauze woven from wire 50 mesh woven from 0.025mm (0.001in)
Tungsten (gold coated)
CAS No.: 7440-33-7 Physical Form: Wire Cloth MDL No.: MFCD00011461
Filtration, electrodes
Vzorec: W
Storage Temperature: Ambient
MDL Number: MFCD00011461
CAS číslo: 7440-33-7
EINECS: 231-143-9