(1S,3R)-(-)-Camphoric acid ≥98%
Dodavatel: Thermo Fisher Scientific
H54042.MD
H54042.03
H54042.06
H54042.MDEA
1310
CZK
H54042.MD
H54042.03
H54042.06
(1S,3R)-(-)-Camphoric acid ≥98%
(1S,3R)-(-)-Camphoric acid
Purity: 98%
CAS No.: 560-09-8
Molecular Formula: C10H16O4
Formula Weight: 200.23
Melting Point: 188-190°
Beilstein Reference: 1874674
Merck Reference: 14,1733
Optical Rotation: -48° (c=10 in ethanol)
MDL No.: MFCD00064937
Vzorec:
C₁₀H₁₆O₄ Bod tání: 188…190 °C Storage Temperature: Ambient |
MDL Number:
MFCD00064937 CAS číslo: 560-09-8 Merck Index: 14,1733 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...