Tisk…
Streptavidin Magnetic Beads
Dodavatel: TRILINK BIOTECHNOLOGIES
M-1002-010 M-1004-010 N-1000-1000 N-1000-010 N-1000-005 N-1000-002 M-1002-100 M-1003-050 M-1003-100 M-1002-020
733-2507EA 26630 CZK
733-2507 SOLUM-1004-010 SOLUN-1000-1000 SOLUN-1000-010 SOLUN-1000-005 SOLUN-1000-002 SOLUM-1002-100 SOLUM-1003-050 SOLUM-1003-100 SOLUM-1002-020
Streptavidin Magnetic Beads
Kuličky Kuličky magnetické
Streptavidin magnetic beads and agarose with biotin-binding capacity as much as 12× higher than comparable products.
Higher binding translates into reduced bead mass or agarose required to immobilise a biotinylated sample and lower background noise from non specific binding, resulting in better signals and lower net costs.