Tisk…

4-Methoxy-3-pyridineboronic acid hydrate 97%

Dodavatel: Thermo Scientific
Varování

Synonyms: (4-Methoxypyridin-3-yl)boronic acid hydrate, 4-Methoxypyridine-3-boronic acid hydrate

378382500 378380010
ACRO378382500EA 2020 CZK
ACRO378382500 ACRO378380010
4-Methoxy-3-pyridineboronic acid hydrate 97%
4-Methoxy-3-pyridineboronic acid hydrate
Vzorec: C₆H₈BNO₃·H₂O
MW: 152,94 g/mol (anhy)
Storage Temperature: Refrigerator
MDL Number: MFCD07186213
CAS číslo: 1266615-79-5

Objednat

Výsledky specifikačního testu

Appearance Cream Powder
Infrared spectrum Conforms
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR