Tisk…

Methyl-3-amino-4-methylbenzoate 97%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: Methyl 3-amino-4-methylbenzoate

A15348.06 A15348.14 A15348.22
A15348.06EA 1250 CZK
A15348.06 A15348.14 A15348.22
Methyl-3-amino-4-methylbenzoate 97%
Methyl-3-amino-4-methylbenzoate

Purity: 97% CAS No.: 18595-18-1 Molecular Formula: C9H11NO2 Formula Weight: 165.19 Melting Point: 113-117° Beilstein Reference: 2088611

Vzorec: C₉H₁₁NO₂
Bod tání: 113…117 °C
Storage Temperature: Ambient
MDL Number: MFCD00025206
CAS číslo: 18595-18-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR