Tisk…

Methyl-3-amino-4-iodobenzoate 97%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: Methyl 3-amino-4-iodobenzoate

B25699.03 B25699.06
B25699.03EA 1490 CZK
B25699.03 B25699.06
Methyl-3-amino-4-iodobenzoate 97%
Methyl-3-amino-4-iodobenzoate

Purity: 97% CAS No.: 412947-54-7 Molecular Formula: C8H8INO2 Formula Weight: 277.06 Melting Point: 134-136° MDL No.: MFCD08064225

Vzorec: C₈H₈INO₂
Bod tání: 134…136 °C
Storage Temperature: Ambient
MDL Number: MFCD08064225
CAS číslo: 412947-54-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR