Tisk…

Methyl-4-amino-3-nitrobenzoate 97%

Dodavatel: Thermo Scientific
Varování

Synonyms: Methyl 4-amino-3-nitrobenzoate

H34348.03 H34348.09
H34348.03EA 1290 CZK
H34348.03 H34348.09
Methyl-4-amino-3-nitrobenzoate 97%
Methyl-4-amino-3-nitrobenzoate

Purity: 97% CAS No.: 3987-92-6 Molecular Formula: C8H8N2O4 Formula Weight: 196.16 Melting Point: 188-191° MDL No.: MFCD00017562

Vzorec: C₈H₈N₂O₄
MW: 196,16 g/mol
Bod tání: 188…191 °C
Storage Temperature: Ambient
MDL Number: MFCD00017562
CAS číslo: 3987-92-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR