2-(Methylthio)benzonitrile ≥98%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
L06870.06
L06870.03
L06870.06EA
6660
CZK
L06870.06
L06870.03
2-(Methylthio)benzonitrile ≥98%
2-(Methylthio)benzonitrile
Purity: 98%
CAS No.: 6609-54-7
Molecular Formula: C8H7NS
Formula Weight: 149.22
Melting Point: 33-38°
Boiling Point: 116-117°/5mm
UN No.: 3439
MDL No.: MFCD00015557
Vzorec:
C₈H₇NS MW: 149,22 g/mol Bod varu: 116…117 °C (5 mmHg) Bod tání: 33…38 °C Storage Temperature: Ambient |
MDL Number:
MFCD00015557 CAS číslo: 6609-54-7 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...