Tisk…

Methyl-2-chloro-6-methoxyisonicotinate ≥98%

Dodavatel: Thermo Scientific
Varování

Synonyms: Methyl 2-chloro-6-methoxypyridine-4-carboxylate, Methyl 2-chloro-6-methoxyisonicotinate

H33438.06 H33438.14
H33438.06EA 3880 CZK
H33438.06 H33438.14
Methyl-2-chloro-6-methoxyisonicotinate ≥98%
Methyl-2-chloro-6-methoxyisonicotinate

Purity: 98% CAS No.: 42521-10-8 Molecular Formula: C8H8ClNO3 Formula Weight: 201.61 Melting Point: 103-105° MDL No.: MFCD00173929

Vzorec: C₈H₈ClNO₃
MW: 201,61 g/mol
Bod tání: 103…105 °C
Storage Temperature: Ambient
MDL Number: MFCD00173929
CAS číslo: 42521-10-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR