Tisk…

Methyl-3,5-dichloroisonicotinate, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Methyl 3,5-dichloropyridine-4-carboxylate

M-Clarity™ quality level = MQ100

679577-1G 679577-5G
SIAL679577-1GEA 0 CZK
SIAL679577-1G SIAL679577-5G
Methyl-3,5-dichloroisonicotinate, Sigma-Aldrich®
Methyl-3,5-dichloroisonicotinate
Vzorec: C₇H₅Cl₂NO₂
MW: 206,026 g/mol
MDL Number: MFCD06203945
CAS číslo: 773874-72-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR