Tisk…

2,6-Dichloroquinoline, Sigma-Aldrich®

Dodavatel: Merck

557803-1G 557803-5G
SIAL557803-1GEA 3380 CZK
SIAL557803-1G SIAL557803-5G
2,6-Dichloroquinoline, Sigma-Aldrich®
2,6-Dichloroquinoline
Vzorec: C₉H₅Cl₂N
MW: 198,0500 g/mol
MDL Number: MFCD03427194
CAS číslo: 1810-72-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR