3-Phenylpropionitrile ≥98%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
Hydrocinnamonitrile
L06483.09
L06483.18
L06483.09EA
1310
CZK
L06483.09
L06483.18
3-Phenylpropionitrile ≥98%
3-Phenylpropionitrile
Purity: 98%
CAS No.: 645-59-0
Molecular Formula: C6H5(CH2)2CN
Formula Weight: 131.18
Flash Point:
Vzorec:
C₉H₉N MW: 131,18 g/mol Bod varu: 261 °C Bod tání: –2 °C Hustota: 1,008 Bod vzplanutí: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001961 CAS číslo: 645-59-0 EINECS: 211-447-8 UN: 3276 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...